(2-Cyanoethyl)hydrazine 95%

Supplier: Ambeed
Danger

Synonyms: 3-Hydrazinylpropanenitrile

A562360-1G A562360-5G A562360-1G
77307-812EA 24 USD
77307-812 103828-750 103839-884
(2-Cyanoethyl)hydrazine 95%
(2-Cyanoethyl)hydrazine

Caution: for Research Use Only

Formula: C₃H₇N₃
MW: 85.11 g/mol
Boiling Pt: 305.6 °C (760 mm Hg)
Flash Pt: 108.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00043620
CAS Number: 353-07-1
UN: 2927
ADR: 6.1,8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR