(3-Fluoro-4-methoxyphenyl)acetic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Fluoro-4-methoxyphenylacetic acid
A176879-1G
A176879-5G
103807-178EA
8
USD
103807-178
103845-506
(3-Fluoro-4-methoxyphenyl)acetic acid 97%
(3-Fluoro-4-methoxyphenyl)acetic acid
Caution: Sealed in dry, at room temperature.
Formula:
C₉H₉FO₃ MW: 184.17 g/mol Boiling Pt: 309 °C (760 mm Hg) Melting Pt: 111…113 °C Flash Pt: 99.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00060353 CAS Number: 452-14-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...