3-Fluoro-2-nitrophenol 98%

Supplier: Ambeed

Warning

A343295-5G A343295-1G
103836-856EA 19 USD
103836-856 103825-882
3-Fluoro-2-nitrophenol 98%
3-Fluoro-2-nitrophenol

Caution: Inert atmosphere, 2…8 °C.

Formula: C₆H₄FNO₃
MW: 157.10 g/mol
Boiling Pt: 237.6 °C (760 mm Hg)
Melting Pt: 37…38 °C
Flash Pt: 97.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07368753
CAS Number: 385-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR