3-Bromo-4-formylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromopyridine-4-carboxaldehyde3-Bromo-4-pyridinecarboxaldehyde3-Bromoisonicotinaldehyde

A110184-1G A110184-250MG A110184-5G
76809-902EA 6.18 USD
76809-902 103823-440 103797-634
3-Bromo-4-formylpyridine 98%
3-Bromo-4-formylpyridine

Caution: for Research Use Only

Formula: C₆H₄BrNO
MW: 186.01 g/mol
Melting Pt: 81…82 °C
Flash Pt: 102.7 °C
Storage Temperature: Ambient
MDL Number: MFCD05864506
CAS Number: 70201-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR