3-Bromo-5-fluorobenzaldehyde 98%

Supplier: Ambeed
Warning

A136746-25G A136746-10G A136746-5G A136746-100G A136746-1G A136746-1G A136746-250MG
77315-268EA 87 USD
77315-268 76767-562 76767-564 77315-266 103802-344 103833-326 76767-566
3-Bromo-5-fluorobenzaldehyde 98%
3-Bromo-5-fluorobenzaldehyde

Caution: for Research Use Only

Formula: C₇H₄BrFO
Boiling Pt: 230.9 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD04116319
CAS Number: 188813-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR