DL-β-Leucine 97%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-4-methylpentanoic acid

A605222-250MG A605222-5G A605222-25G A605222-1G
77117-294EA 220.42 USD
77117-294 77117-292 77117-290 103828-998
DL-β-Leucine 97%
DL-β-Leucine

Caution: for Research Use Only

Formula: C₆H₁₃NO₂
MW: 131.1729 g/mol
Boiling Pt: 232 °C (760 mm Hg)
Melting Pt: 206…209 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00800505
CAS Number: 5699-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR