3-Amino-4-methoxypyridine 98%

Supplier: Ambeed
Warning

A166798-1G A166798-250MG A166798-1G A166798-100MG A166798-5G
77163-370EA 7 USD
77163-370 77320-642 77163-368 103844-360 103806-098
3-Amino-4-methoxypyridine 98%
3-Amino-4-methoxypyridine

Caution: Keep in a dark place, inert atmosphere, store in freezer, under -20 °C

Formula: C₆H₈N₂O
MW: 124.1400 g/mol
Melting Pt: 60…66 °C
Flash Pt: 117.4 °C
Storage Temperature: Freezer
MDL Number: MFCD07374959
CAS Number: 33631-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR