Hexaethylene glycol 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3,6,9,12,15-Pentaoxaheptadecane-1,17-diol
A195221-1G
A195221-100G
A195221-5G
A195221-10G
A195221-5G
77085-296EA
21.63
USD
77085-296
76800-158
76800-160
103816-730
103847-344
Hexaethylene glycol 97%
Hexaethylene glycol
Caution: for Research Use Only
Formula:
C₁₂H₂₆O₇ MW: 282.33 g/mol Boiling Pt: 217 °C (760 mm Hg) Melting Pt: 5…7 °C Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002877 CAS Number: 2615-15-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...