3,4,5-Trifluoroiodobenzene 98%
Supplier: Ambeed
|
Warning
|
A108039-1G
A108039-5G
A108039-1G
A108039-5G
A108039-25G
A108039-100G
77270-684EA
149
USD
77270-684
103797-284
103823-058
77309-900
77270-682
3,4,5-Trifluoroiodobenzene 98%
3,4,5-Trifluoroiodobenzene
Caution: for Research Use Only
Formula:
C₆H₂F₃I MW: 257.9800 g/mol Boiling Pt: 176.4 °C (760 mm Hg) Flash Pt: 65.1 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03265259 CAS Number: 170112-66-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...