3-(3,4-Dimethylphenyl)propionic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-(3,4-Dimethylphenyl)propanoic acid

A100405-1G A100405-5G A100405-250MG
76994-442EA 12.36 USD
76994-442 103821-118 103795-422
3-(3,4-Dimethylphenyl)propionic acid 98%
3-(3,4-Dimethylphenyl)propionic acid

Caution: for Research Use Only

Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
Boiling Pt: 311.4 °C (760 mm Hg)
Melting Pt: 80…82 °C
Flash Pt: 208.5 °C
Storage Temperature: Ambient
MDL Number: MFCD03425686
CAS Number: 25173-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR