2-Methyl-6-nitrobenzoic anhydride 97%
Supplier: Ambeed
|
Warning
|
A209783-5G
A209783-5G
A209783-25G
A209783-1G
A209783-1G
A209783-250MG
103848-644EA
48.46
USD
103848-644
77086-998
77087-000
77086-996
103817-958
76873-636
2-Methyl-6-nitrobenzoic anhydride 97%
2-Methyl-6-nitrobenzoic anhydride
Caution: for Research Use Only
Formula:
C₁₆H₁₂N₂O₇ MW: 344.2800 g/mol Melting Pt: 177 °C Flash Pt: 246.455 °C Storage Temperature: Ambient |
MDL Number:
MFCD06797117 CAS Number: 434935-69-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...