2-Methoxyquinoline-3-boronic acid 97%

Supplier: Ambeed
Warning

A114189-1G A114189-100MG A114189-250MG A114189-5G A114189-5G A114189-1G
77311-022EA 502 USD
77311-022 103798-364 77311-020 77025-966 77025-964 103824-204
2-Methoxyquinoline-3-boronic acid 97%
2-Methoxyquinoline-3-boronic acid

Caution: for Research Use Only

Formula: C₁₀H₁₀BNO₃
MW: 203.0000 g/mol
Boiling Pt: 414.2 °C (760 mm Hg)
Flash Pt: 204.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09800894
CAS Number: 886853-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR