2-Isopropoxy-5-pyridineboronic acid pinacol ester 98%

Warning

Synonyms: 6-Isopropoxy-3-pyridylboronic acid Pinacol ester2-Isopropoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine2-Isopropoxypyridine-5-boronic acid pinacol ester2-(6-Isopropoxy-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A128909-1G A128909-5G
103801-200EA 41.54 USD
103801-200 103832-082
2-Isopropoxy-5-pyridineboronic acid pinacol ester 98%
2-Isopropoxy-5-pyridineboronic acid pinacol ester

Caution: for Research Use Only

Formula: C₁₄H₂₂BNO₃
MW: 263.14 g/mol
Boiling Pt: 347.6 °C (760 mm Hg)
Melting Pt: 72…75 °C
Flash Pt: 164 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07368871
CAS Number: 871839-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR