2-Iodo-5-methoxybenzoic acid 97%

Supplier: Ambeed

Warning

A208219-5G A208219-1G
103817-862EA 10 USD
103817-862 103848-550
2-Iodo-5-methoxybenzoic acid 97%
2-Iodo-5-methoxybenzoic acid

Caution: Keep in a dark place, sealed in dry, room temperature

Formula: C₈H₇IO₃
MW: 278.0400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00204590
CAS Number: 54413-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR