4-Methylsalicylaldehyde 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Hydroxy-4-methylbenzaldehyde
A152935-1G
A152935-1G
A152935-10G
A152935-25G
A152935-5G
A152935-250MG
A152935-5G
77318-254EA
6
USD
77318-254
103842-622
77318-258
76689-436
77318-256
103804-452
76689-438
4-Methylsalicylaldehyde 98%
4-Methylsalicylaldehyde
Caution: for Research Use Only
Formula:
C₈H₈O₂ MW: 136.15 g/mol Boiling Pt: 219.5 °C (760 mm Hg) Melting Pt: 58…61 °C Storage Temperature: Ambient |
MDL Number:
MFCD00799550 CAS Number: 698-27-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...