2H-Pyrido[3,2-b][1,4]oxazin-3(4H)-one ≥97%
Supplier: Ambeed
|
Warning
|
A120892-1G
A120892-5G
103800-006EA
16
USD
103800-006
103830-820
2H-Pyrido[3,2-b][1,4]oxazin-3(4H)-one ≥97%
2H-Pyrido[3,2-b][1,4]oxazin-3(4H)-one
Caution: Sealed in dry, at room temperature.
Formula:
C₇H₆N₂O₂ Boiling Pt: 384.2 °C (760 mm Hg) Melting Pt: 204…208 °C Flash Pt: 186.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006697 CAS Number: 20348-09-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...