2-Fluoro-3-methylbenzaldehyde 98%

Supplier: AMBEED, INC

Warning

A376439-1G A376439-25G A376439-5G
76694-390EA 177 USD
76694-390 103826-582 103837-608
2-Fluoro-3-methylbenzaldehyde 98%
2-Fluoro-3-methylbenzaldehyde

Caution: for Research Use Only

Formula: C₈H₇FO
MW: 138.1390 g/mol
Boiling Pt: 197.4 °C (760 mm Hg)
Flash Pt: 79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06797948
CAS Number: 886762-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR