2'-Deoxycytidine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one
, deoxyC
A106243-1G
A106243-5G
103822-768EA
19
USD
103822-768
103797-016
2'-Deoxycytidine 97%
2'-Deoxycytidine
Caution: Keep in a dark place, sealed in dry, store in freezer, under –20°C
Formula:
C₉H₁₃N₃O₄ MW: 227.22 g/mol Boiling Pt: 497.6 °C (760 mm Hg) Melting Pt: 209…211 °C Flash Pt: 254.8 °C Storage Temperature: Freezer |
MDL Number:
MFCD00006547 CAS Number: 951-77-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...