2-Chloroquinoxaline 98%

Supplier: Ambeed
Warning

A160115-250MG A160115-1G A160115-5G A160115-1G A160115-100G A160115-5G A160115-25G A160115-10G
103805-334EA 6.92 USD
103805-334 103843-564 77319-484 77259-576 77319-486 77319-488 77259-572 77259-574
2-Chloroquinoxaline 98%
2-Chloroquinoxaline

Caution: for Research Use Only

Formula: C₈H₅ClN₂
MW: 164.5917 g/mol
Boiling Pt: 252.9 °C (760 mm Hg)
Melting Pt: 47…50 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00043907
CAS Number: 1448-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR