2-Chloro-3-fluorobenzaldehyde 95%
Supplier: Ambeed
|
Warning
|
A142475-100G
A142475-5G
A142475-25G
A142475-10G
A142475-1G
A142475-5G
A142475-1G
77299-300EA
22
USD
77299-300
76767-870
103803-164
76767-872
103834-196
77299-296
77299-298
2-Chloro-3-fluorobenzaldehyde 95%
2-Chloro-3-fluorobenzaldehyde
Caution: for Research Use Only
Formula:
C₇H₄ClFO MW: 158.56 g/mol Boiling Pt: 211 °C (760 mm Hg) Flash Pt: 81.407 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD07782053 CAS Number: 96516-31-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...