6-Bromosalicylaldehyde 97%

Supplier: Ambeed

Warning

Synonyms: 2-Bromo-6-hydroxybenzaldehyde

A122998-1G A122998-5G
103800-324EA 14 USD
103800-324 103831-160
6-Bromosalicylaldehyde 97%
6-Bromosalicylaldehyde

Caution: Inert atmosphere, 2…8 °C.

Formula: C₇H₅BrO₂
MW: 201.0200 g/mol
Boiling Pt: 243.4 °C (760 mm Hg)
Flash Pt: 101.001 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08234722
CAS Number: 22532-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR