2-Bromo-5-nitropyridine 98%
Supplier: Ambeed
|
Warning
|
A241768-10G
A241768-5G
A241768-1G
A241768-5G
A241768-25G
77276-022EA
12
USD
77276-022
103851-232
77304-222
77276-024
77090-394
2-Bromo-5-nitropyridine 98%
2-Bromo-5-nitropyridine
Caution: for Research Use Only
Formula:
C₅H₃BrN₂O₂ MW: 203 g/mol Boiling Pt: 289.3 °C (760 mm Hg) Melting Pt: 139…141 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006222 CAS Number: 4487-59-6 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...