2-Bromo-4-(trifluoromethoxy)benzonitrile 97%

Supplier: Ambeed

Warning

A408561-1G A408561-5G
103838-236EA 139 USD
103838-236 103827-190
2-Bromo-4-(trifluoromethoxy)benzonitrile 97%
2-Bromo-4-(trifluoromethoxy)benzonitrile

Caution: Sealed in dry, at room temperature.

Formula: C₈H₃BrF₃NO
MW: 266.0147 g/mol
Boiling Pt: 265 °C (760 mm Hg)
Flash Pt: 114 °C
Storage Temperature: Ambient
MDL Number: MFCD13185874
CAS Number: 1214334-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR