2-Bromo-3-methylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-3-picoline

A179455-5G A179455-100G A179455-25G A179455-5G A179455-500G A179455-1G A179455-10G
77287-124EA 180 USD
77287-124 77083-236 77287-122 77287-120 77083-238 103845-778 77287-118
2-Bromo-3-methylpyridine 98%
2-Bromo-3-methylpyridine

Caution: for Research Use Only

Formula: C₆H₆BrN
MW: 172.02 g/mol
Boiling Pt: 215.3 °C (760 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00239380
CAS Number: 3430-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR