2-Bromo-3-chloroaniline 98%
Supplier: Ambeed
|
Warning
|
A192835-100G
A192835-5G
A192835-10G
A192835-25G
A192835-1G
A192835-5G
A192835-250MG
A192835-1G
A192835-500G
76784-994EA
1823.1
USD
76784-994
103847-094
77084-980
76784-992
103816-484
77084-976
77084-978
76784-988
76784-990
2-Bromo-3-chloroaniline 98%
2-Bromo-3-chloroaniline
Caution: for Research Use Only
Formula:
C₆H₅BrClN MW: 206.4700 g/mol Boiling Pt: 277 °C (760 mm Hg) Melting Pt: 41…43 °C Flash Pt: 122 °C Storage Temperature: Ambient |
MDL Number:
MFCD10699543 CAS Number: 96558-73-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...