2-Bromo-4'-fluoro-3'-(trifluoromethyl)acetophenone 97%

Supplier: Ambeed
Danger

Synonyms: 2-Bromo-1-(4-fluoro-3-(trifluoromethyl)phenyl)ethanone4-Fluoro-3-(trifluoromethyl)phenacyl bromide

A342057-1G A342057-10G A342057-5G
103836-832EA 38.08 USD
103836-832 77290-546 103825-858
2-Bromo-4'-fluoro-3'-(trifluoromethyl)acetophenone 97%
2-Bromo-4'-fluoro-3'-(trifluoromethyl)acetophenone

Caution: for Research Use Only

Formula: C₉H₅BrF₄O
MW: 285.0330 g/mol
Boiling Pt: 249.7 °C (760 mm Hg)
Melting Pt: 42…44 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03094509
CAS Number: 537050-14-9
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR