2-Aminoterephthalic acid 98%
Supplier: Ambeed
|
Warning
|
A145885-100G
A145885-500G
A145885-25G
A145885-5G
A145885-5G
77235-192EA
525
USD
77235-192
77316-992
77316-990
103841-684
77235-190
2-Aminoterephthalic acid 98%
2-Aminoterephthalic acid
Caution: for Research Use Only
Formula:
C₈H₇NO₄ Melting Pt: 324 °C Flash Pt: 226.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00134536 CAS Number: 10312-55-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...