2-Amino-5-fluoro-6-methylpyridine 96%
Supplier: Ambeed
|
Warning
|
A112131-5G
A112131-25G
A112131-100MG
A112131-1G
A112131-5G
77270-970EA
469
USD
77270-970
103797-958
77270-968
103823-792
77284-210
2-Amino-5-fluoro-6-methylpyridine 96%
2-Amino-5-fluoro-6-methylpyridine
Caution: for Research Use Only
Formula:
C₆H₇FN₂ MW: 126.13 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09835298 CAS Number: 110919-71-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...