2-Amino-5-bromonicotinaldehyde 98%

Supplier: Ambeed

Warning

A164674-5G A164674-1G
103844-146EA 185 USD
103844-146 103805-888
2-Amino-5-bromonicotinaldehyde 98%
2-Amino-5-bromonicotinaldehyde

Caution: Keep in a dark place, inert atmosphere, 2…8 °C

Formula: C₆H₅BrN₂O
Melting Pt: 167…169 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09607904
CAS Number: 206997-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR