4-Iodoanthranilic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Amino-4-iodobenzoic acid
A100118-1G
A100118-10G
A100118-100G
A100118-250MG
A100118-25G
A100118-5G
A100118-1G
A100118-5G
77245-058EA
630
USD
77245-058
103795-260
77282-434
77296-576
77245-052
77245-054
103820-980
77245-056
4-Iodoanthranilic acid 97%
4-Iodoanthranilic acid
Caution: for Research Use Only
Formula:
C₇H₆INO₂ MW: 263.0325 g/mol Boiling Pt: 377 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD09800365 CAS Number: 20776-54-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...