3-Iodoanthranilic acid 97%

Supplier: Ambeed

Warning

Synonyms: 2-Amino-3-iodobenzoic acid

A111829-5G A111829-1G
103797-910EA 52 USD
103797-910 103823-732
3-Iodoanthranilic acid 97%
3-Iodoanthranilic acid

Caution: Keep in a dark place, inert atmosphere, room temperature

Formula: C₇H₆INO₂
MW: 263.0300 g/mol
Boiling Pt: 366.5 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD06808542
CAS Number: 20776-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR