2-Amino-3,5-dibromopyrazine 98%

Supplier: Ambeed

Warning

A844496-5G A844496-1000G
103810-584EA 522 USD
103810-584 103841-214
2-Amino-3,5-dibromopyrazine 98%
2-Amino-3,5-dibromopyrazine

Caution: Keep in a dark place, inert atmosphere, 2…8 °C

Formula: C₄H₃Br₂N₃
MW: 252.90 g/mol
Boiling Pt: 294.6 °C (760 mm Hg)
Melting Pt: 114…117 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00673150
CAS Number: 24241-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR