2,8-Dichloroquinazoline 97%

Supplier: Ambeed

Warning

A158671-1G A158671-5G
103843-366EA 760 USD
103843-366 103805-150
2,8-Dichloroquinazoline 97%
2,8-Dichloroquinazoline

Caution: Inert atmosphere, 2…8 °C.

Formula: C₈H₄Cl₂N₂
Storage Temperature: Refrigerator
MDL Number: MFCD13193682
CAS Number: 67092-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR