2,6-Dichloroisonicotinonitrile 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Cyano-2,6-dichloropyridine
A376568-25G
A376568-1G
A376568-250MG
A376568-5G
A376568-1G
A376568-5G
A376568-100G
103826-584EA
8.08
USD
103826-584
77155-266
76840-914
77306-880
77306-882
77155-268
103837-610
2,6-Dichloroisonicotinonitrile 98%
2,6-Dichloroisonicotinonitrile 2-(4-Formylphenyl)isonicotinonitrile
Caution: for Research Use Only
Formula:
C₆H₂Cl₂N₂ MW: 172.9995 g/mol Boiling Pt: 239.4 °C (760 mm Hg) Melting Pt: 96 °C Flash Pt: 98.6 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00173784 CAS Number: 32710-65-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...