4,6-Difluoro-m-toluic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,4-Difluoro-5-methylbenzoic acid
A104874-1G
A104874-5G
103822-518EA
154
USD
103822-518
103796-792
4,6-Difluoro-m-toluic acid 97%
4,6-Difluoro-m-toluic acid
Caution: Sealed in dry, at room temperature.
Formula:
C₈H₆F₂O₂ MW: 172.13 g/mol Melting Pt: 159…166 °C Storage Temperature: Ambient |
MDL Number:
MFCD09835285 CAS Number: 367954-99-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...