2,4-Dichloroquinazoline ≥97%

Supplier: Ambeed

Warning

A144334-1G A144334-5G
103841-468EA 15 USD
103841-468 103803-390
2,4-Dichloroquinazoline ≥97%
2,4-Dichloroquinazoline

Caution: Inert atmosphere, store in freezer, under –20 °C.

Formula: C₈H₄Cl₂N₂
Boiling Pt: 302.9 °C (760 mm Hg)
Melting Pt: 121 °C at N/A mmHg
Flash Pt: 137 °C
Storage Temperature: Freezer
MDL Number: MFCD00091950
CAS Number: 607-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR