2,4-Dichloro-7-methoxyquinazoline 95%
Supplier: Ambeed
|
Warning
|
A188742-5G
A188742-100MG
A188742-1G
A188742-5G
A188742-250MG
77302-150EA
470
USD
77302-150
77248-492
103846-722
77248-494
103816-130
2,4-Dichloro-7-methoxyquinazoline 95%
2,4-Dichloro-7-methoxyquinazoline
Caution: for Research Use Only
Formula:
C₉H₆Cl₂N₂O MW: 229.0600 g/mol Boiling Pt: 244.4 °C (760 mm Hg) Flash Pt: 101.6 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09954881 CAS Number: 62484-31-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...