2,4,6-Trichloroquinazoline 97%

Supplier: Ambeed
Warning

A253498-250MG A253498-1G A253498-5G A253498-5G
77091-378EA 299.73 USD
77091-378 103851-944 103812-094 77249-984
2,4,6-Trichloroquinazoline 97%
2,4,6-Trichloroquinazoline

Caution: for Research Use Only

Formula: C₈H₃Cl₃N₂
MW: 233.4818 g/mol
Boiling Pt: 298.5 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD08445622
CAS Number: 20028-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR