2,3-Dimethyl-4-nitrophenol 97%
|
Warning
|
A101033-1G
A101033-5G
A101033-10G
103795-768EA
10.38
USD
103795-768
103821-460
2,3-Dimethyl-4-nitrophenol 97%
2,3-Dimethyl-4-nitrophenol
Caution: for Research Use Only
Formula:
C₈H₉NO₃ MW: 167.1600 g/mol Boiling Pt: 323.9 °C (760 mm Hg) Flash Pt: 145.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD01861284 CAS Number: 19499-93-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...