Tetrafluoroterephthalic acid 97%

Supplier: Ambeed

Warning

Synonyms: 2,3,5,6-Tetrafluoroterephthalic acid

A235500-1G A235500-5G
103850-786EA 7 USD
103850-786 103811-018
Tetrafluoroterephthalic acid 97%
Tetrafluoroterephthalic acid

Caution: Sealed in dry, at room temperature.

Formula: C₈H₂F₄O₄
MW: 238.0900 g/mol
Melting Pt: 275…277 °C
Flash Pt: 158.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00012301
CAS Number: 652-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR