2-(4-Methylpiperazino)-3-pyridinamine 97%

Supplier: Ambeed

Warning

Synonyms: 2-(4-Methylpiperazin-1-yl)pyridin-3-amine

A197944-5G A197944-1G
103847-604EA 465 USD
103847-604 103816-978
2-(4-Methylpiperazino)-3-pyridinamine 97%
2-(4-Methylpiperazino)-3-pyridinamine

Caution: Keep in a dark place, inert atmosphere, 2…8 °C

Formula: C₁₀H₁₆N₄
MW: 192.27 g/mol
Melting Pt: 112…114 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01704018
CAS Number: 5028-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR