4-Chloro-3-fluorophenylacetonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Chloro-3-fluorophenyl)acetonitrile

A187530-5G A187530-10G A187530-1G A187530-1G A187530-25G
103846-612EA 36.92 USD
103846-612 77302-088 77084-308 103816-022 76998-890
4-Chloro-3-fluorophenylacetonitrile 98%
4-Chloro-3-fluorophenylacetonitrile

Caution: for Research Use Only

Formula: C₈H₅ClFN
MW: 169.58 g/mol
Boiling Pt: 253.4 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD04115860
CAS Number: 251570-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR