(4,5-Difluoro-2-nitrophenyl)acetic acid 97%

Supplier: Ambeed

Warning

Synonyms: 2-(4,5-Difluoro-2-nitrophenyl)acetic acid

A718929-5G
103840-728EA 350 USD
103840-728
(4,5-Difluoro-2-nitrophenyl)acetic acid 97%
(4,5-Difluoro-2-nitrophenyl)acetic acid

Caution: Sealed in dry, at room temperature.

Formula: C₈H₅F₂NO₄
MW: 217.1300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09878318
CAS Number: 1000339-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR