1-Ethylnaphthalene ≥95%

Supplier: Ambeed

Warning

A115867-1G A115867-5G
103824-502EA 19 USD
103824-502 103798-640
1-Ethylnaphthalene ≥95%
1-Ethylnaphthalene

Caution: Sealed in dry, at room temperature.

Formula: C₁₂H₁₂
MW: 156.23 g/mol
Boiling Pt: 259.1 °C (760 mm Hg)
Melting Pt: …15 °C
Flash Pt: 111.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00004049
CAS Number: 1127-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR