1-Cyanocyclopropanecarboxylic acid 97%

Supplier: Ambeed

Danger

Synonyms: 1-Cyano-1-cyclopropanecarboxylic acid

A122118-1G A122118-5G
103831-004EA 27 USD
103831-004 103800-178
1-Cyanocyclopropanecarboxylic acid 97%
1-Cyanocyclopropanecarboxylic acid

Caution: Sealed in dry, at 2…8°C

Formula: C₅H₅NO₂
MW: 111.1 g/mol
Melting Pt: 140 °C
Flash Pt: 150.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00190651
CAS Number: 6914-79-0
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR