1-Chloroisoquinoline ≥97%

Supplier: Ambeed

Warning

A211376-1G A211376-5G
103818-100EA 6 USD
103818-100 103848-794
1-Chloroisoquinoline ≥97%
1-Chloroisoquinoline

Caution: Inert atmosphere, 2…8 °C.

Formula: C₉H₆ClN
MW: 163.61 g/mol
Boiling Pt: 274.5 °C (760 mm Hg)
Melting Pt: 31…36 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00024134
CAS Number: 19493-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR