1-Bromooctadecane 95%

Supplier: Ambeed
Warning

A835369-100G A835369-5G A835369-25G
77244-410EA 8 USD
77244-410 103841-186 77308-430
1-Bromooctadecane 95%
1-Bromooctadecane

Caution: for Research Use Only

Formula: BrH₂C(CH₂)₁₆CH₃
MW: 333.4 g/mol
Boiling Pt: 370.2 °C (760 mm Hg)
Melting Pt: 25…30 °C at N/A mmHg
Flash Pt: 150.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00000231
CAS Number: 112-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR