Bromopentamethylbenzene 98%

Supplier: Ambeed

Warning

Synonyms: Pentamethylbromobenzene1-Bromo-2,3,4,5,6-pentamethylbenzene

A175374-5G A175374-1G
103807-034EA 8 USD
103807-034 103845-340
Bromopentamethylbenzene 98%
Bromopentamethylbenzene

Caution: Sealed in dry, at room temperature.

Formula: C₁₁H₁₅Br
MW: 227.14 g/mol
Boiling Pt: 286.5 °C (760 mm Hg)
Melting Pt: 161…162 °C
Flash Pt: 126 °C
Storage Temperature: Ambient
MDL Number: MFCD00013523
CAS Number: 5153-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR