1,4-Dichloroisoquinoline 97%
Supplier: Ambeed
|
Warning
|
A130676-1G
A130676-250MG
A130676-10G
A130676-5G
103801-456EA
19.62
USD
103801-456
77094-952
103832-358
77094-950
1,4-Dichloroisoquinoline 97%
1,4-Dichloroisoquinoline
Caution: for Research Use Only
Formula:
C₉H₅Cl₂N MW: 198.0500 g/mol Melting Pt: 92…94 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD05664199 CAS Number: 15298-58-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...