α,α'-Dibromo-p-xylene 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
p-Xylylene dibromide
, 1,4-Bis(bromomethyl)benzene
A575067-5G
103839-974EA
21
USD
103839-974
α,α'-Dibromo-p-xylene 98%
α,α'-Dibromo-p-xylene
Caution: Keep in a dark place, sealed in dry, room temperature
Formula:
C₈H₈Br₂ MW: 263.96 g/mol Boiling Pt: 245 °C (760 mm Hg) Melting Pt: 143…145 °C Flash Pt: 118.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000182 CAS Number: 623-24-5 UN: 2928 ADR: 6.1,8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...